siRNA Details
Gene Symbol: D730047P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021347.1 | - | 1 | 466 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730940.1 | - | 1 | 462 | Chr. 9 - 78821880 - 78822371 [+] on Build GRCm38 |
