siRNA Details
Gene Symbol: D730047E02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021345.1 | - | 2 | 492 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730934.1 | - | 2 | 489 | Chr. 12 - 55390361 - 55391499 [-] on Build GRCm38 |
