siRNA Details
Gene Symbol: 1700085B13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007003.1 | - | 321 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707059.1 | - | 1 | 314 | Chr. 7 - 28436611 - 28436954 [+] on Build GRCm38 |
