siRNA Details
Gene Symbol: Gm20743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040303.1 | 2 | 436 | |||
GenBank mRNA | AK034030.1 | - | 200 | |||
AK042927.1 | - | 2 | 425 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm20743 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040303.1 | 2 | 436 | |||
GenBank mRNA | AK034030.1 | - | 200 | |||
AK042927.1 | - | 2 | 425 |