siRNA Details
Gene Symbol: Gm41505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142264.1 | - | 680 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB477468.2 | - | 1 | 140 | Chr. 16 - 94749243 - 94753438 [+] on Build GRCm38 |
