siRNA Details
Gene Symbol: Gm41505 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142264.1 | - | 425 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB659173.1 | - | 2 | 425 | Chr. 16 - 94721894 - 94749326 [+] on Build GRCm38 |
