siRNA Details
Gene Symbol: Gm34240 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085621.1 | - | 2 | 283 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB661610.1 | - | 2 | 283 | Chr. 3 - 63439251 - 63456598 [+] on Build GRCm38 |
