siRNA Details
Gene Symbol: 5330439A09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019937.1 | - | 460 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717046.1 | - | 2 | 459 | Chr. 8 - 54569053 - 54600987 [-] on Build GRCm38 |
