siRNA Details
Gene Symbol: 5330439K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019939.1 | - | 2 | 554 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY717050.1 | - | 2 | 547 | Chr. 15 - 100461351 - 100468685 [-] on Build GRCm38 |
