siRNA Details
Gene Symbol: Nrip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078829.1 | - | 1 | 610 | ||
AK088511.1 | - | 1 | 2850 | |||
BC062260.1 | - | 1 | 301 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CK031677.1 | - | 1 | 626 | Chr. 16 - 76287934 - 76288662 [-] on Build GRCm38 |
