siRNA Details
Gene Symbol: 9230114J08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK020347.1 | - | 146 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721158.1 | - | 1 | 142 | Chr. 3 - 96998712 - 97000768 [-] on Build GRCm38 |
