siRNA Details
Gene Symbol: 1700081N11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006973.1 | - | 2 | 479 | ||
AV205735.1 | - | 1 | 285 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707027.1 | - | 2 | 479 | Chr. 12 - 54862991 - 54863664 [+] on Build GRCm38 |
