siRNA Details
Gene Symbol: 1700021F13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006202.1 | - | 1 | 89 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706297.1 | - | 1 | 89 | Chr. 5 - 119805547 - 119807869 [-] on Build GRCm38 |
