siRNA Details
Gene Symbol: 1700021J08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006209.1 | - | 2 | 359 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706301.1 | - | 2 | 359 | Chr. 4 - 117002166 - 117003012 [-] on Build GRCm38 |
