siRNA Details
Gene Symbol: 2010110E17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008378.1 | - | 1 | 239 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708292.1 | - | 1 | 239 | Chr. 8 - 86885688 - 86886541 [+] on Build GRCm38 |
