siRNA Details
Gene Symbol: Gm16907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045794.1 | 1 | 91 | |||
GenBank mRNA | AK030196.1 | - | 1 | 92 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB617085.1 | - | 1 | 89 | Chr. 13 - 63291307 - 63297145 [-] on Build GRCm38 |
