siRNA Details
Gene Symbol: 2210414L08Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019066.1 | - | 1 | 288 | ||
AK131897.1 | - | 1 | 288 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708814.1 | - | 1 | 288 | Chr. 18 - 67968192 - 67968867 [+] on Build GRCm38 |
