siRNA Details
Gene Symbol: Thrb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC042730.1 | - | 12 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BG865050.1 | - | 72 | Chr. 14 - 17813271 - 18036967 [+] on Build GRCm38 |
