siRNA Details
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033718.1 | - | 1 | 2857 | ||
AK083275.1 | - | 2195 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Traf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK033718.1 | - | 1 | 2857 | ||
AK083275.1 | - | 2195 |