siRNA Details
Gene Symbol: Gm8327 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB030205.1 | - | 1 | 455 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AI450677.1 | - | 1 | 440 | Chr. 13 - 78199285 - 78199758 [+] on Build GRCm38 |
