siRNA Details
Gene Symbol: Gm2912 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK148097.1 | - | 433 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY741294.1 | - | 4 | 433 | Chr. 12 - 68970570 - 68988333 [+] on Build GRCm38 |
