siRNA Details
Gene Symbol: Pantr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC094545.1 | - | 409 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BU054879.1 | - | 4 | 410 | Chr. 1 - 42591995 - 42694808 [-] on Build GRCm38 |
