siRNA Details
Gene Symbol: 2700057C20Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK012449.1 | - | 1 | 476 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY711583.1 | - | 1 | 474 | Chr. 7 - 12498752 - 12499646 [+] on Build GRCm38 |
