siRNA Details
Gene Symbol: Gm36173 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK136704.1 | - | 2 | 392 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY721096.1 | - | 2 | 382 | Chr. 10 - 53235187 - 53260247 [+] on Build GRCm38 |
