siRNA Details
Gene Symbol: 1810073G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK007969.1 | - | 1 | 1652 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | CF585213.1 | - | 1 | 555 | Chr. 19 - 58743150 - 58743775 [+] on Build GRCm38 |
