siRNA Details
Gene Symbol: 1700003P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045982.1 | 1, 2 | 44 | |||
GenBank mRNA | AK005657.1 | - | 1, 2 | 45 | ||
AK006568.1 | - | 1, 2 | 60 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 1700003P14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045982.1 | 1, 2 | 44 | |||
GenBank mRNA | AK005657.1 | - | 1, 2 | 45 | ||
AK006568.1 | - | 1, 2 | 60 |