siRNA Details
Gene Symbol: Gm3161 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK090034.1 | - | 364 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY733074.1 | - | 2 | 363 | Chr. 14 - 43275034 - 43289107 [+] on Build GRCm38 |
