siRNA Details
Gene Symbol: Gm26579 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK144843.1 | - | 1 | 270 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY734490.1 | - | 1 | 270 | Chr. 10 - 116674591 - 116679064 [+] on Build GRCm38 |
