siRNA Details
Gene Symbol: 1700045H11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040649.1 | 4 | 419 | |||
GenBank mRNA | AK006698.1 | - | 4 | 421 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706690.1 | - | 4 | 417 | Chr. 4 - 150828223 - 150854940 [-] on Build GRCm38 |
