siRNA Details
Gene Symbol: 1700042O13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK006691.1 | - | 1 | 152 | ||
AK030060.1 | - | 1 | 148 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY706682.1 | - | 1 | 149 | Chr. 3 - 8743680 - 8744557 [+] on Build GRCm38 |
