siRNA Details
Gene Symbol: D530014G21Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021287.1 | - | 2 | 446 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730571.1 | - | 2 | 443 | Chr. 8 - 8488304 - 8495535 [+] on Build GRCm38 |
