siRNA Details
Gene Symbol: 5830434F19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017969.1 | - | 1 | 1376 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV316091.1 | - | 1 | 86 | Chr. 2 - 34417029 - 34417275 [+] on Build GRCm38 |
