siRNA Details
Gene Symbol: C030039E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK021134.1 | - | 1 | 303 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY607232.1 | - | 1 | 16 | Chr. 13 - 29015743 - 29016122 [+] on Build GRCm38 |
