siRNA Details
Gene Symbol: Gm36607 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK039809.1 | - | 4 | 528 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB198567.2 | - | 2 | 61 | Chr. 13 - 70707975 - 70733207 [+] on Build GRCm38 |
