siRNA Details
Gene Symbol: 2010300F17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK008491.1 | - | 2 | 644 | ||
AK145005.1 | - | 1 | 1048 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY708382.1 | - | 2 | 644 | Chr. 11 - 96746116 - 96747410 [-] on Build GRCm38 |
