siRNA Details
Gene Symbol: 1110036E04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040713.1 | 2 | 161 | |||
GenBank mRNA | AK004124.1 | - | 2 | 160 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY704366.1 | - | 2 | 156 | Chr. 9 - 64049833 - 64054100 [+] on Build GRCm38 |
