siRNA Details
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142440.1 | - | 432 | |||
AK165789.1 | - | 1305 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY730563.1 | - | 2 | 432 | Chr. 6 - 36387402 - 36388917 [-] on Build GRCm38 |
