siRNA Details
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142440.1 | - | 627 | |||
AK165789.1 | - | 1501 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 9330158H04Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK142440.1 | - | 627 | |||
AK165789.1 | - | 1501 |