siRNA Details
Gene Symbol: 4833412C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045954.1 | 3 | 406 | |||
GenBank mRNA | AK014683.1 | - | 3 | 407 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4833412C05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045954.1 | 3 | 406 | |||
GenBank mRNA | AK014683.1 | - | 3 | 407 |