siRNA Details
Gene Symbol: 4833412E19Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK014685.1 | - | 1 | 1368 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB692744.1 | - | 1 | 152 | Chr. 6 - 120009359 - 120009835 [-] on Build GRCm38 |
