siRNA Details
Gene Symbol: Gm32568 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK089350.1 | - | 3 | 1202 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY764325.1 | - | 1 | 487 | Chr. 8 - 66496254 - 66496946 [+] on Build GRCm38 |
