siRNA Details
Gene Symbol: Fam198b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK132613.1 | - | 246 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB614161.1 | - | 1 | 246 | Chr. 3 - 79886158 - 79886806 [-] on Build GRCm38 |
