siRNA Details
Gene Symbol: 4930519K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019684.1 | - | 1, 2 | 142 | ||
BY715389.1 | - | 1, 2 | 142 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: 4930519K11Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019684.1 | - | 1, 2 | 142 | ||
BY715389.1 | - | 1, 2 | 142 |