siRNA Details
Gene Symbol: 4930520M14Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK019689.1 | - | 2 | 505 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715405.1 | - | 2 | 505 | Chr. 5 - 139215495 - 139216356 [-] on Build GRCm38 |
