siRNA Details
Gene Symbol: 2900046L07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK013658.1 | - | 1 | 1719 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | AV152916.1 | - | 1 | 132 | Chr. 15 - 44716448 - 44716654 [-] on Build GRCm38 |
