siRNA Details
Gene Symbol: 4930415P13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015156.1 | - | 1 | 373 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY714703.1 | - | 1 | 373 | Chr. 18 - 46339896 - 46340429 [-] on Build GRCm38 |
