siRNA Details
Gene Symbol: 4930558K02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK016179.1 | - | 1 | 85 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY715782.1 | - | 1 | 87 | Chr. 1 - 161942282 - 161969141 [-] on Build GRCm38 |
