siRNA Details
Gene Symbol: 5830400J07Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK017886.1 | - | 1 | 1200 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BX517703.1 | - | 1 | 482 | Chr. 12 - 80865546 - 80866054 [+] on Build GRCm38 |
