siRNA Details
Gene Symbol: 1810021B22Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040417.1 | 1 | 208 | |||
GenBank mRNA | AK007571.1 | - | 1 | 209 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BY707618.1 | - | 1 | 206 | Chr. 15 - 89071196 - 89075852 [-] on Build GRCm38 |
