siRNA Details
Gene Symbol: Gm13783 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038769.1 | - | 1 | 21 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BB632042.1 | - | 1 | 21 | Chr. 2 - 91195995 - 91202778 [-] on Build GRCm38 |
